Assay Details
Gene Symbol: JHDM1D-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_024451.1 | 1 | 147 | 106 | ||
GenBank mRNA | DA585317.1 | - | 1 | 147 | 106 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | lysine demethylase 7A | KDM7A | JHDM1D | -305.0 | Anti-sense | Chr. 7 - 140084746 - 140176941 on NCBI Build GRCh38 |
Downstream | solute carrier family 37 member 3 | SLC37A3 | 154.1kb | Anti-sense | Chr. 7 - 140333752 - 140404352 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07000878.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 618135 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P54503 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P54503 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 24835 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 54262 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P54503 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_32_P54503 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05826735 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26085293 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26085272 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26085277 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26085268 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26085275 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16024467 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05826746 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26085282 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26085276 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26085294 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05826751 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26085270 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26085290 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05826754 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05826759 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05826756 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05826741 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26085289 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05826749 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05826748 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05826752 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26085275 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26085277 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05826735 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05826759 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16024467 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05826749 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05826741 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05826746 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26085282 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26085293 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05826756 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26085272 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05826748 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26085270 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26085268 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05826754 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05826751 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26085294 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26085290 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26085276 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05826752 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26085289 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_32_P54503 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P54503 | - | - |
Agilent Human 2 cDNA Microarray (G4101A) | 3225132 | - | - |
Agilent Human 2 cDNA Microarray (G4101A) | 3225132 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P136395 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P54503 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05826740 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16024461 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05826733 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05826767 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05826760 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05826751 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05826758 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05826745 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05826762 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16024468 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05826747 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05826749 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05826742 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05826735 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05826736 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05826753 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05826765 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05826738 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05826756 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05826769 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05826744 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P01821569 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 232579_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0700009360.hg.1 | ENST00000566699.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700009360.hg.1 | NR_024451.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07013732.hg.1 | NR_024451.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07013732.hg.1 | TC07000878.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0700166454.hg.1 | ENST00000566699.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700166454.hg.1 | NR_024451.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700166454.hg.1 | TC0700009360.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700166454.hg.1 | NR_024451.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700166454.hg.1 | ENST00000566699.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009360.hg.1 | ENST00000566699.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700009360.hg.1 | NR_024451.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 232579_at | - | - |
Human Genome U133 Plus 2.0 Array | 232579_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232579_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232579_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232579_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232579_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232579_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232579_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232579_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232579_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232579_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232579_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232579_at5 | - | - |
Human Genome U95 Set | 77039_PM_at | - | - |
Human Genome U95 Set | 72649_PM_at | - | - |
Human Genome U95 Set | 77039_at | - | - |
Human Genome U95 Set | 72649_at | - | - |
Human Genome U133 Set | 232579_at | - | - |
Human Genome U133 Set | 232579_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A035170 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A035170 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3246279 | AK026372.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3246279 | NR_024451.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3246279 | AK026372.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3246279 | NR_024451.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3246279 | AK026372.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3246279 | NR_024451.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3246279 | AK026372.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3246279 | NR_024451.1 | human_transcriptome |
