Assay Details
Gene Symbol: CLCA3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_024604.1 | 14 - 15 | 2437 | 81 | ||
GenBank mRNA | AF043976.1 | - | 14 - 15 | 2438 | 81 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | chloride channel accessory 4 | CLCA4 |
CaCC, CaCC2 |
-69.4kb | Sense | Chr. 1 - 86547076 - 86580758 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC105378828 | LOC105378828 | 48.1kb | Anti-sense | Chr. 1 - 86571181 - 86704493 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000825.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
transport
ion transmembrane transport
regulation of ion transmembrane transport
calcium ion transmembrane transport
chloride transmembrane transport
metalloendopeptidase activity
transporter activity
voltage-gated ion channel activity
chloride channel activity
calcium channel activity
proteolysis
transport
ion transmembrane transport
regulation of ion transmembrane transport
calcium ion transmembrane transport
chloride transmembrane transport
metalloendopeptidase activity
transporter activity
voltage-gated ion channel activity
chloride channel activity
calcium channel activity
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01013137.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01013137.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01013137.hg.1 | TC01000825.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100163701.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100163701.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100163701.hg.1 | TC0100008925.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100163701.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100163701.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000284054.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008925.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100163700.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100163700.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100163700.hg.1 | NR_024604.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100163700.hg.1 | TC0100008925.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100163700.hg.1 | ENST00000466454.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100163700.hg.1 | ENST00000490028.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100163700.hg.1 | NR_024604.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 106806 | - | - |
