Assay Details
Gene Symbol: HECTD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF086006.1 | - | 1 | 446 | 102 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 6861 | MIR6861 | hsa-mir-6861 | -97.4kb | Sense | Chr. 12 - 112163258 - 112163321 on NCBI Build GRCh38 |
Downstream | ribosomal protein L6 | RPL6 |
L6, SHUJUN-2, TAXREB107, TXREB1 |
143.8kb | Sense | Chr. 12 - 112405181 - 112418850 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
