Assay Details
Gene Symbol: LOC100507437 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125385.1 | 1 - 2 | 64 | |||
GenBank mRNA | AK123017.1 | - | 1 - 2 | 722 | 64 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | testis expressed 22 | TEX22 | -3.7kb | Anti-sense | Chr. 14 - 105398499 - 105413859 on NCBI Build GRCh38 | |
Downstream | metastasis associated 1 | MTA1 | 110.0 | Anti-sense | Chr. 14 - 105419849 - 105470729 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
