Assay Details
Gene Symbol: KRBOX1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_122033.1 | 1 - 2 | 1028 | 114 | ||
GenBank mRNA | AK057338.1 | - | 1 - 2 | 1028 | 114 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | KRAB box domain containing 1 | KRBOX1 | Anti-sense | Chr. 3 - 42936342 - 42942792 on NCBI Build GRCh38 | ||
Downstream | family with sequence similarity 198 member A | FAM198A | C3orf41 | 42.5kb | Anti-sense | Chr. 3 - 42979267 - 43057715 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
