Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013241.hg.1 |
ENST00000455966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013241.hg.1 |
ENST00000455966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013241.hg.1 |
ENST00000455966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013241.hg.1 |
ENST00000455966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013241.hg.1 |
ENST00000455966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013241.hg.1 |
NR_110692.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013241.hg.1 |
NR_110692.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013241.hg.1 |
NR_110692.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013241.hg.1 |
NR_110692.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013241.hg.1 |
NR_110692.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013241.hg.1 |
NR_110692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198230.hg.1 |
ENST00000455966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198230.hg.1 |
NR_110692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198230.hg.1 |
TC0100013241.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100198230.hg.1 |
NR_110692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198230.hg.1 |
ENST00000455966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198238.hg.1 |
NR_110692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198238.hg.1 |
TC0100013241.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100198238.hg.1 |
NR_110692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013241.hg.1 |
ENST00000455966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013241.hg.1 |
ENST00000455966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013241.hg.1 |
ENST00000455966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013241.hg.1 |
ENST00000455966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013241.hg.1 |
ENST00000455966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013241.hg.1 |
NR_110692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013241.hg.1 |
NR_110692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013241.hg.1 |
NR_110692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013241.hg.1 |
NR_110692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013241.hg.1 |
NR_110692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013241.hg.1 |
NR_110692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198229.hg.1 |
ENST00000455966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198229.hg.1 |
NR_110692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198229.hg.1 |
TC0100013241.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100198229.hg.1 |
ENST00000455966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198229.hg.1 |
NR_110692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198228.hg.1 |
ENST00000455966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198228.hg.1 |
NR_110692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198228.hg.1 |
TC0100013241.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100198228.hg.1 |
ENST00000455966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198228.hg.1 |
NR_110692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198233.hg.1 |
ENST00000455966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198233.hg.1 |
NR_110692.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198233.hg.1 |
TC0100013241.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100198233.hg.1 |
ENST00000455966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198233.hg.1 |
NR_110692.1 |
Refseq |