Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010692.hg.1 |
ENST00000319121.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010692.hg.1 |
ENST00000319121.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010692.hg.1 |
ENST00000319121.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010692.hg.1 |
ENST00000319121.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010692.hg.1 |
ENST00000319121.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010692.hg.1 |
ENST00000560400.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010692.hg.1 |
NM_018143.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010692.hg.1 |
NM_018143.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010692.hg.1 |
NM_018143.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010692.hg.1 |
NM_018143.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010692.hg.1 |
NM_018143.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010692.hg.1 |
NM_018143.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010692.hg.1 |
NM_018143.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010692.hg.1 |
ENST00000319121.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010692.hg.1 |
ENST00000319121.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010692.hg.1 |
ENST00000319121.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010692.hg.1 |
ENST00000319121.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010692.hg.1 |
ENST00000319121.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010692.hg.1 |
ENST00000560400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010692.hg.1 |
NM_018143.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010692.hg.1 |
NM_018143.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010692.hg.1 |
NM_018143.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010692.hg.1 |
NM_018143.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010692.hg.1 |
NM_018143.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010692.hg.1 |
NM_018143.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010692.hg.1 |
NM_018143.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700184762.hg.1 |
ENST00000560400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184762.hg.1 |
NM_018143.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700184762.hg.1 |
ENST00000560400.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184762.hg.1 |
NM_018143.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700184762.hg.1 |
TC1700010692.hg.1 |
Clariom D TC |