Assay Details
Gene Symbol: SENP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172109.1 | NP_001165580.1 | 2 - 3 | 443 | 294 | |
NM_001172110.1 | NP_001165581.1 | 2 - 3 | 285 | 190 | ||
XM_011521215.2 | XP_011519517.1 | 2 - 3 | 576 | 190 | ||
GenBank mRNA | BP370384.1 | - | 2 - 3 | 285 | 190 | |
DB048641.1 | - | 2 - 3 | 443 | 294 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC15000654.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR15005605.hg.1 | ENST00000544411.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15005605.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15005605.hg.1 | TC15000654.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR15005606.hg.1 | ENST00000544411.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15005606.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15005606.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15005606.hg.1 | TC15000654.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15005607.hg.1 | ENST00000544411.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15005607.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15005607.hg.1 | TC15000654.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000340912.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000542035.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001166340.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_001172111.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007777.hg.1 | NM_145204.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155200.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155200.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155200.hg.1 | TC1500007777.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1500155200.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155200.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155198.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155198.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155198.hg.1 | TC1500007777.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1500155198.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155198.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155199.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155199.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155199.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155199.hg.1 | NM_001172110.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155199.hg.1 | TC1500007777.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1500155199.hg.1 | ENST00000564863.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155199.hg.1 | ENST00000567794.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500155199.hg.1 | NM_001172109.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500155199.hg.1 | NM_001172110.1 | Refseq |
