Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023343.hg.1 |
ENST00000368636.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023343.hg.1 |
ENST00000540159.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023343.hg.1 |
NM_004052.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10023343.hg.1 |
TC10001769.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000368636.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000368636.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000368636.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000368636.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000368636.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000368636.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000368636.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000368636.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000368636.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000368636.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000368636.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000368636.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000368636.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000540159.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000540159.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000540159.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000540159.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000540159.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000540159.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000540159.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000540159.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000540159.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000540159.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000540159.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000540159.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000631806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000631806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000631806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000631806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000631806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000633835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000633835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000633835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000633835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000633835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000633835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000633835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
ENST00000633835.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
NM_004052.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
NM_004052.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
NM_004052.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
NM_004052.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
NM_004052.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
NM_004052.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
NM_004052.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
NM_004052.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
NM_004052.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
NM_004052.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
NM_004052.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012329.hg.1 |
NM_004052.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000184067.hg.1 |
ENST00000368636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000184067.hg.1 |
ENST00000540159.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000184067.hg.1 |
ENST00000633835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000184067.hg.1 |
NM_004052.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000184067.hg.1 |
TC1000012329.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000184067.hg.1 |
NM_004052.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000184067.hg.1 |
ENST00000368636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000184067.hg.1 |
ENST00000540159.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000184067.hg.1 |
ENST00000633835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000368636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000368636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000368636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000368636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000368636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000368636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000368636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000368636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000368636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000368636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000368636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000368636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000368636.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000540159.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000540159.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000540159.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000540159.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000540159.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000540159.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000540159.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000540159.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000540159.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000540159.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000540159.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000540159.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000631806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000631806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000631806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000631806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000631806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000633835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000633835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000633835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000633835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000633835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000633835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000633835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
ENST00000633835.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
NM_004052.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
NM_004052.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
NM_004052.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
NM_004052.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
NM_004052.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
NM_004052.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
NM_004052.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
NM_004052.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
NM_004052.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
NM_004052.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
NM_004052.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012329.hg.1 |
NM_004052.3 |
Refseq |