Assay Details
Gene Symbol: MIR148B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_029894.1 | 1 | 49 | 92 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | coatomer protein complex subunit zeta 1 | COPZ1 |
CGI-120, COPZ, HSPC181, zeta-COP, zeta1-COP |
Sense | Chr. 12 - 54325090 - 54351851 on NCBI Build GRCh38 | |
Downstream | G protein-coupled receptor 84 | GPR84 |
EX33, GPCR4 |
13.5kb | Anti-sense | Chr. 12 - 54362445 - 54365224 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200007760.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12000463.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364157 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364160 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28799499 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28799475 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364172 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364137 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364161 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364143 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364132 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28799471 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28799505 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364144 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364134 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364139 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364148 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364164 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364163 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28799498 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28799488 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16561098 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364135 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28799479 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364153 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28799503 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364151 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28799485 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28799506 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28799495 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28799506 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28799475 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364172 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364144 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28799503 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364139 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28799505 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364161 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28799498 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28799479 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28799485 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364132 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28799471 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16561098 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364153 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364134 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28799495 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364160 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364164 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364163 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364135 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364148 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364143 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364137 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364157 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28799499 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28799488 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08364151 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.1 ST Array | 7955906 | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC1200007760.hg.1 | ENST00000362252.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007760.hg.1 | NR_029894.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12006031.hg.1 | ENST00000362252.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12006031.hg.1 | NR_029894.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12006031.hg.1 | TC12000463.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1200155774.hg.1 | ENST00000362252.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200155774.hg.1 | NR_029894.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200155774.hg.1 | TC1200007760.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200155774.hg.1 | NR_029894.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200155774.hg.1 | ENST00000362252.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007760.hg.1 | ENST00000362252.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007760.hg.1 | NR_029894.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3308260 | NR_029894.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3308260 | NR_029894.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3308260 | NR_029894.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3308260 | NR_029894.1 | human_transcriptome |
