Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06026681.hg.1 |
NR_037938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06026681.hg.1 |
TC06002125.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06026682.hg.1 |
NR_037938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06026682.hg.1 |
TC06002125.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013229.hg.1 |
ENST00000406908.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013229.hg.1 |
NR_037938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013229.hg.1 |
NR_037938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013229.hg.1 |
NR_037938.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013229.hg.1 |
NR_037938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013229.hg.1 |
ENST00000406908.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013229.hg.1 |
NR_037938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013229.hg.1 |
NR_037938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013229.hg.1 |
NR_037938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013229.hg.1 |
NR_037938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191323.hg.1 |
NR_037938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191323.hg.1 |
TC0600013229.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600191323.hg.1 |
NR_037938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191325.hg.1 |
NR_037938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191325.hg.1 |
TC0600013229.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600191325.hg.1 |
NR_037938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191326.hg.1 |
NR_037938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191326.hg.1 |
TC0600013229.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600191326.hg.1 |
NR_037938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191324.hg.1 |
ENST00000406908.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191324.hg.1 |
NR_037938.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191324.hg.1 |
TC0600013229.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600191324.hg.1 |
ENST00000406908.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191324.hg.1 |
NR_037938.1 |
Refseq |