Assay Details
Gene Symbol: BAG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204878.1 | NP_001191807.1 | 2 - 3 | 807 | 103 | |
NM_004874.3 | NP_004865.1 | 3 - 4 | 915 | 103 | ||
GenBank mRNA | AF095194.2 | - | 3 - 4 | 675 | 103 | |
AF111116.1 | - | 3 - 4 | 633 | 103 | ||
AK304072.1 | - | 2 - 3 | 584 | 103 | ||
AK314020.1 | - | 3 - 4 | 688 | 103 | ||
BC000800.1 | - | 2 - 3 | 807 | 103 | ||
BC038505.1 | - | 3 - 4 | 709 | 5259771 | 103 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08000287.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
protein folding
positive regulation of fibroblast migration
positive regulation of actin filament polymerization
positive regulation of peptidyl-serine phosphorylation
tumor necrosis factor-mediated signaling pathway
negative regulation of apoptotic process
positive regulation of cell adhesion
positive regulation of stress fiber assembly
positive regulation of protein kinase B signaling
cellular response to tumor necrosis factor
cellular response to epidermal growth factor stimulus
protein localization to plasma membrane
ruffle assembly
regulation of cellular response to heat
negative regulation of protein targeting to mitochondrion
negative regulation of phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase activity
adenyl-nucleotide exchange factor activity
RNA binding
protein binding
ubiquitin protein ligase binding
chaperone binding
protein folding
positive regulation of fibroblast migration
positive regulation of actin filament polymerization
positive regulation of peptidyl-serine phosphorylation
tumor necrosis factor-mediated signaling pathway
negative regulation of apoptotic process
positive regulation of cell adhesion
positive regulation of stress fiber assembly
positive regulation of protein kinase B signaling
cellular response to tumor necrosis factor
cellular response to epidermal growth factor stimulus
protein localization to plasma membrane
ruffle assembly
regulation of cellular response to heat
negative regulation of protein targeting to mitochondrion
negative regulation of phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase activity
adenyl-nucleotide exchange factor activity
RNA binding
protein binding
ubiquitin protein ligase binding
chaperone binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR08003973.hg.1 | ENST00000287322.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08003973.hg.1 | ENST00000432471.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08003973.hg.1 | NM_001204878.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08003973.hg.1 | NM_004874.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08003973.hg.1 | TC08000287.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000287322.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000287322.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000287322.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000287322.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000287322.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000287322.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000287322.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000287322.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000287322.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000287322.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000287322.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000287322.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000432471.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000432471.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000432471.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000432471.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000432471.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000432471.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000432471.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000432471.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000432471.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000432471.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000521282.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000521282.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000521282.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000521282.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000521282.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000521311.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000521311.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000521311.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | ENST00000521311.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | NM_001204878.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | NM_001204878.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | NM_001204878.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | NM_001204878.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | NM_001204878.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | NM_001204878.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | NM_001204878.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | NM_001204878.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | NM_001204878.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | NM_001204878.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | NM_001204878.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | NM_004874.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | NM_004874.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | NM_004874.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | NM_004874.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | NM_004874.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | NM_004874.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | NM_004874.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | NM_004874.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | NM_004874.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | NM_004874.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | NM_004874.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007328.hg.1 | NM_004874.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000287322.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000287322.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000287322.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000287322.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000287322.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000287322.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000287322.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000287322.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000287322.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000287322.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000287322.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000287322.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000432471.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000432471.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000432471.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000432471.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000432471.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000432471.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000432471.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000432471.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000432471.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000432471.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000521282.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000521282.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000521282.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000521282.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000521282.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000521311.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000521311.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000521311.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | ENST00000521311.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | NM_001204878.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | NM_001204878.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | NM_001204878.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | NM_001204878.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | NM_001204878.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | NM_001204878.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | NM_001204878.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | NM_001204878.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | NM_001204878.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | NM_001204878.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | NM_001204878.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | NM_004874.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | NM_004874.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | NM_004874.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | NM_004874.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | NM_004874.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | NM_004874.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | NM_004874.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | NM_004874.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | NM_004874.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | NM_004874.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | NM_004874.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007328.hg.1 | NM_004874.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800150341.hg.1 | ENST00000287322.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150341.hg.1 | ENST00000432471.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150341.hg.1 | ENST00000521311.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150341.hg.1 | NM_001204878.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800150341.hg.1 | NM_004874.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800150341.hg.1 | TC0800007328.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800150341.hg.1 | NM_001204878.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800150341.hg.1 | ENST00000287322.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150341.hg.1 | ENST00000432471.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150341.hg.1 | ENST00000521311.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150341.hg.1 | NM_004874.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800150340.hg.1 | ENST00000287322.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150340.hg.1 | ENST00000432471.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150340.hg.1 | ENST00000521311.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150340.hg.1 | NM_001204878.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800150340.hg.1 | NM_004874.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800150340.hg.1 | TC0800007328.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800150340.hg.1 | ENST00000287322.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150340.hg.1 | ENST00000432471.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150340.hg.1 | ENST00000521311.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800150340.hg.1 | NM_001204878.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800150340.hg.1 | NM_004874.3 | Refseq |
