Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016446.hg.1 |
ENST00000429044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016446.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016446.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016446.hg.1 |
TC09001318.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016447.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016447.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016447.hg.1 |
TC09001318.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016443.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016443.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016443.hg.1 |
TC09001318.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016444.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016444.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016444.hg.1 |
TC09001318.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
ENST00000427523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
ENST00000427523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
ENST00000427523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
ENST00000429044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
ENST00000429044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173098.hg.1 |
ENST00000429044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173098.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173098.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173098.hg.1 |
TC0900010736.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900173098.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173098.hg.1 |
ENST00000429044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173098.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
ENST00000427523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
ENST00000427523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
ENST00000427523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
ENST00000429044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
ENST00000429044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173093.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173093.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173093.hg.1 |
TC0900010736.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900173093.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173093.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173099.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173099.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173099.hg.1 |
TC0900010736.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900173099.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173099.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173095.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173095.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173095.hg.1 |
TC0900010736.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900173095.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173095.hg.1 |
NR_034157.1 |
Refseq |