Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
ENST00000427523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
ENST00000427523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
ENST00000427523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
ENST00000429044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
ENST00000429044.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016441.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016441.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016441.hg.1 |
TC09001318.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016442.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016442.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016442.hg.1 |
TC09001318.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0900173091.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173091.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173091.hg.1 |
TC0900010736.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900173091.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173091.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173092.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173092.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173092.hg.1 |
TC0900010736.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900173092.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173092.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
ENST00000427523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
ENST00000427523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
ENST00000427523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
ENST00000429044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
ENST00000429044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
ENST00000449990.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010736.hg.1 |
NR_034157.1 |
Refseq |