Assay Details
Gene Symbol: A2MP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040112.1 | 3 - 4 | 592 | 107 | ||
GenBank mRNA | DY655604.1 | - | 3 - 4 | 107 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
PZP, alpha-2-macroglobulin like |
PZP | CPAMD6 | -23.7kb | Sense | Chr. 12 - 9140730 - 9208370 on NCBI Build GRCh38 |
Downstream | microRNA 1244-3 | MIR1244-3 | 6.9kb | Sense | Chr. 12 - 9239467 - 9239551 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001189.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR12015826.hg.1 | ENST00000543404.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12015826.hg.1 | NR_040112.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12015826.hg.1 | TC12001189.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000543404.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000543404.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000543404.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000543404.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000543404.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000543404.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000543404.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000543404.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000543404.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000543404.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000544183.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000544183.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000544183.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000544183.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000544183.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000544183.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000544183.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000545343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000545343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000545343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000545343.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | NR_040112.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | NR_040112.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | NR_040112.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | NR_040112.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | NR_040112.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | NR_040112.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | NR_040112.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | NR_040112.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | NR_040112.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009854.hg.1 | NR_040112.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200174190.hg.1 | ENST00000543404.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200174190.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200174190.hg.1 | NR_040112.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200174190.hg.1 | TC1200009854.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200174190.hg.1 | NR_040112.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200174190.hg.1 | ENST00000543404.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200174190.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000543404.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000543404.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000543404.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000543404.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000543404.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000543404.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000543404.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000543404.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000543404.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000543404.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000544183.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000544183.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000544183.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000544183.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000544183.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000544183.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000544183.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000545343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000545343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000545343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000545343.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | ENST00000566278.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | NR_040112.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | NR_040112.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | NR_040112.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | NR_040112.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | NR_040112.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | NR_040112.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | NR_040112.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | NR_040112.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | NR_040112.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009854.hg.1 | NR_040112.1 | Refseq |
