Assay Details
Gene Symbol: LOC100506321 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045122.1 | 3 | 826 | 110 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | CHURC1-FNTB readthrough | CHURC1-FNTB | -31.0kb | Sense | Chr. 14 - 64914361 - 65062655 on NCBI Build GRCh38 | |
Downstream | uncharacterized LOC100128233 | LOC100128233 | 118.6kb | Anti-sense | Chr. 14 - 65212893 - 65222347 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC14000396.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | ENST00000555261.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | ENST00000555261.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | ENST00000555261.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | ENST00000555261.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | ENST00000555898.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | ENST00000555898.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | ENST00000555898.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | ENST00000555898.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | ENST00000555898.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | ENST00000555898.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | ENST00000555898.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | ENST00000555898.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | ENST00000556127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | ENST00000556127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007458.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14004582.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14004582.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14004582.hg.1 | TC14000396.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR14004591.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14004591.hg.1 | ENST00000555261.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14004591.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14004591.hg.1 | TC14000396.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR14004593.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14004593.hg.1 | TC14000396.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR14004588.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14004588.hg.1 | ENST00000555261.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14004588.hg.1 | ENST00000555898.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14004588.hg.1 | ENST00000556127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14004588.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14004588.hg.1 | TC14000396.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR14004590.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14004590.hg.1 | ENST00000555261.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14004590.hg.1 | ENST00000555898.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14004590.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14004590.hg.1 | TC14000396.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR14004592.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14004592.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14004592.hg.1 | TC14000396.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1400152168.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152168.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152168.hg.1 | ENST00000555261.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152168.hg.1 | ENST00000555898.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152168.hg.1 | ENST00000555898.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152168.hg.1 | ENST00000556127.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152168.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400152168.hg.1 | TC1400007458.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400152168.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152168.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152168.hg.1 | ENST00000555261.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152168.hg.1 | ENST00000555898.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152168.hg.1 | ENST00000555898.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152168.hg.1 | ENST00000556127.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152168.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | ENST00000555261.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | ENST00000555261.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | ENST00000555261.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | ENST00000555261.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | ENST00000555898.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | ENST00000555898.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | ENST00000555898.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | ENST00000555898.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | ENST00000555898.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | ENST00000555898.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | ENST00000555898.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | ENST00000555898.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | ENST00000556127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | ENST00000556127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007458.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400152171.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152171.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152171.hg.1 | ENST00000555261.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152171.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400152171.hg.1 | TC1400007458.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400152171.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152171.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152171.hg.1 | ENST00000555261.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152171.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400152170.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152170.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152170.hg.1 | ENST00000555261.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152170.hg.1 | ENST00000555898.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152170.hg.1 | ENST00000555898.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152170.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400152170.hg.1 | TC1400007458.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400152170.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152170.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152170.hg.1 | ENST00000555261.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152170.hg.1 | ENST00000555898.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152170.hg.1 | ENST00000555898.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152170.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400152172.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152172.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152172.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400152172.hg.1 | TC1400007458.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400152172.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152172.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152172.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400152162.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152162.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152162.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400152162.hg.1 | TC1400007458.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400152162.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152162.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152162.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400152173.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152173.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152173.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400152173.hg.1 | TC1400007458.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400152173.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152173.hg.1 | ENST00000553633.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400152173.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400152174.hg.1 | NR_045122.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400152174.hg.1 | TC1400007458.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400152174.hg.1 | NR_045122.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 201013 | - | - |
