Assay Details
Gene Symbol: LOC646626 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045484.1 | 2 | 725 | 107 | ||
GenBank mRNA | BX104049.1 | - | 2 | 330 | 105 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | B-cell CLL/lymphoma 10 | BCL10 |
CARMEN, CIPER, CLAP, IMD37, c-E10, mE10 |
-737.0 | Anti-sense | Chr. 1 - 85265776 - 85276904 on NCBI Build GRCh38 |
Downstream | dimethylarginine dimethylaminohydrolase 1 | DDAH1 |
DDAH, HEL-S-16 |
40.4kb | Anti-sense | Chr. 1 - 85318485 - 85578363 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
