Assay Details
Gene Symbol: MIR5702 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_049887.1 | 1 | 44 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC646736 | LOC646736 | -478.6kb | Anti-sense | Chr. 2 - 226142794 - 226180062 on NCBI Build GRCh38 | |
Downstream | insulin receptor substrate 1 | IRS1 | HIRS-1 | 72.5kb | Sense | Chr. 2 - 226731317 - 226798790 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
