Assay Details
Gene Symbol: MAX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271068.1 | NP_001257997.1 | 2 - 3 | 344 | 112 | |
NM_145114.2 | NP_660089.1 | 3 - 4 | 371 | 112 | ||
NR_073138.1 | 2 - 3 | 290 | 112 | |||
GenBank mRNA | AI769390.1 | - | 1 - 2 | 112 | ||
BC013669.1 | - | 3 - 4 | 336 | 4155088 | 112 | |
HY018878.1 | - | 2 - 3 | 217 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
