Assay Details
Gene Symbol: PPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000310.3 | NP_000301.1 | 7 - 8 | 957 | 94 | |
NM_001142604.1 | NP_001136076.1 | 4 - 5 | 648 | 94 | ||
GenBank mRNA | AK302232.1 | - | 4 - 5 | 430 | 94 | |
AK310799.1 | - | 7 - 8 | 809 | 94 | ||
AK312287.1 | - | 7 - 8 | 732 | 94 | ||
BC008426.1 | - | 7 - 8 | 737 | 4249991 | 94 | |
CR542053.1 | - | 7 - 8 | 725 | 94 | ||
U44772.1 | - | 7 - 8 | 738 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
