Assay Details
Gene Symbol: TFG-ALK fusion | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF125093.1 | - | 456 | 77 | ||
AF143407.1 | - | 456 | 242 | |||
OM_TFG-ALK_T4A20_COSF424.0 | - | 56 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
