Assay Details
Gene Symbol: EML4-ALK fusion | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB374364.1 | - | 1 | 292 | 112 | |
AB374365.1 | - | 1 | 409 | 229 | ||
JQ828841.1 | - | 1 | 408 | 295 | ||
OM_EML4-ALK_E2A20_COSF478.0 | - | 73 | 86 | |||
OM_EML4-ALK_E2A20_COSF478_1.0 | - | 99 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
