Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
ENST00000567665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
ENST00000567665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
ENST00000567665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
ENST00000567665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
ENST00000567665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
ENST00000568751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
ENST00000568751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
ENST00000568751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
ENST00000568751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
ENST00000568751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
ENST00000568751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
ENST00000568751.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
ENST00000569164.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
ENST00000569164.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
ENST00000569164.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
NR_104660.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
NR_104660.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
NR_104660.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
NR_104660.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
NR_104660.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
NR_104660.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
NR_104660.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011435.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191605.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191605.hg.1 |
TC1600011435.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600191605.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191600.hg.1 |
ENST00000568751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191600.hg.1 |
NR_104660.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191600.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191600.hg.1 |
TC1600011435.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600191600.hg.1 |
ENST00000568751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191600.hg.1 |
NR_104660.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191600.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191607.hg.1 |
ENST00000567665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191607.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191607.hg.1 |
TC1600011435.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600191607.hg.1 |
ENST00000567665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191607.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191601.hg.1 |
ENST00000567665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191601.hg.1 |
ENST00000568751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191601.hg.1 |
NR_104660.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191601.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191601.hg.1 |
TC1600011435.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600191601.hg.1 |
ENST00000567665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191601.hg.1 |
ENST00000568751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191601.hg.1 |
NR_104660.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191601.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191604.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191604.hg.1 |
TC1600011435.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600191604.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
ENST00000567665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
ENST00000567665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
ENST00000567665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
ENST00000567665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
ENST00000567665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
ENST00000568751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
ENST00000568751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
ENST00000568751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
ENST00000568751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
ENST00000568751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
ENST00000568751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
ENST00000568751.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
ENST00000569164.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
ENST00000569164.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
ENST00000569164.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
NR_104660.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
NR_104660.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
NR_104660.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
NR_104660.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
NR_104660.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
NR_104660.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
NR_104660.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011435.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191608.hg.1 |
ENST00000567665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191608.hg.1 |
NR_104661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191608.hg.1 |
TC1600011435.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600191608.hg.1 |
ENST00000567665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191608.hg.1 |
NR_104661.1 |
Refseq |