Assay Details
Gene Symbol: CTB-7E3.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_108020.1 | 2 - 3 | 495 | 109 | ||
GenBank mRNA | DB052051.1 | - | 2 - 3 | 496 | 109 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC102546299 | LOC102546299 | -2.4mb | Anti-sense | Chr. 5 - 164470279 - 164542984 on NCBI Build GRCh38 | |
Downstream | teneurin transmembrane protein 2 | TENM2 |
ODZ2, TEN-M2, TNM2, ten-2 |
71.1kb | Anti-sense | Chr. 5 - 166979066 - 168264159 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
