Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009445.hg.1 |
ENST00000557871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009445.hg.1 |
ENST00000557871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009445.hg.1 |
ENST00000557871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009445.hg.1 |
ENST00000557871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009445.hg.1 |
ENST00000557871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009445.hg.1 |
ENST00000557871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009445.hg.1 |
ENST00000557871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009445.hg.1 |
ENST00000557871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009445.hg.1 |
ENST00000557871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009445.hg.1 |
ENST00000557871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009445.hg.1 |
NR_109775.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009445.hg.1 |
NR_109775.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009445.hg.1 |
NR_109775.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009445.hg.1 |
NR_109775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500169382.hg.1 |
ENST00000557871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500169382.hg.1 |
NR_109775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500169382.hg.1 |
TC1500009445.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500169382.hg.1 |
NR_109775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500169382.hg.1 |
ENST00000557871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009445.hg.1 |
ENST00000557871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009445.hg.1 |
ENST00000557871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009445.hg.1 |
ENST00000557871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009445.hg.1 |
ENST00000557871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009445.hg.1 |
ENST00000557871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009445.hg.1 |
ENST00000557871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009445.hg.1 |
ENST00000557871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009445.hg.1 |
ENST00000557871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009445.hg.1 |
ENST00000557871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009445.hg.1 |
ENST00000557871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009445.hg.1 |
NR_109775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009445.hg.1 |
NR_109775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009445.hg.1 |
NR_109775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009445.hg.1 |
NR_109775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500169383.hg.1 |
ENST00000557871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500169383.hg.1 |
NR_109775.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500169383.hg.1 |
TC1500009445.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500169383.hg.1 |
ENST00000557871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500169383.hg.1 |
NR_109775.1 |
Refseq |