Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000621402.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000621402.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000622928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000622928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000624367.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000624367.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000624367.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000624367.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000624367.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000624367.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000624367.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000624367.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000624367.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000624385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000624385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000624385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000624385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000624385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000624385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
ENST00000624385.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
NR_109880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
NR_109880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
NR_109880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
NR_109880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
NR_109880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
NR_109880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
NR_109880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006909.hg.1 |
NR_109880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000147304.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147304.hg.1 |
ENST00000624367.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147304.hg.1 |
ENST00000624385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147304.hg.1 |
NR_109880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000147304.hg.1 |
TC2000006909.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000147304.hg.1 |
NR_109880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000147304.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147304.hg.1 |
ENST00000624367.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000147304.hg.1 |
ENST00000624385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000621402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000621402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000622928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000622928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000623256.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000624367.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000624367.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000624367.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000624367.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000624367.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000624367.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000624367.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000624367.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000624367.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000624385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000624385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000624385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000624385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000624385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000624385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
ENST00000624385.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
NR_109880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
NR_109880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
NR_109880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
NR_109880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
NR_109880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
NR_109880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
NR_109880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006909.hg.1 |
NR_109880.1 |
Refseq |