Assay Details
Gene Symbol: LOC101927292 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110048.1 | 3 - 4 | 2624 | 67 | ||
GenBank mRNA | BC034605.1 | - | 3 - 4 | 2624 | 67 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC283332 | LOC283332 | -11.9kb | Anti-sense | Chr. 12 - 49908882 - 49911863 on NCBI Build GRCh38 | |
Downstream | aquaporin 2 | AQP2 |
AQP-CD, WCH-CD |
25.7kb | Sense | Chr. 12 - 49950741 - 49958881 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000547443.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000547443.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000547443.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000547443.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000547443.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000547443.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000547954.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000547954.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000547954.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000547954.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000547954.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000547954.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000547954.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000549734.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000549734.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000551539.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000551539.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | ENST00000551539.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200007588.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200153555.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153555.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153555.hg.1 | ENST00000547954.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153555.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200153555.hg.1 | TC1200007588.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200153555.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200153555.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153555.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153555.hg.1 | ENST00000547954.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153557.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153557.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153557.hg.1 | ENST00000547954.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153557.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200153557.hg.1 | TC1200007588.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200153557.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153557.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153557.hg.1 | ENST00000547954.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153557.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200153558.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153558.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153558.hg.1 | ENST00000547443.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153558.hg.1 | ENST00000547954.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153558.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200153558.hg.1 | TC1200007588.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200153558.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153558.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153558.hg.1 | ENST00000547443.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153558.hg.1 | ENST00000547954.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153558.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000547443.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000547443.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000547443.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000547443.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000547443.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000547443.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000547954.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000547954.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000547954.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000547954.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000547954.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000547954.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000547954.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000549734.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000549734.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000551539.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000551539.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | ENST00000551539.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200007588.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200153553.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153553.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153553.hg.1 | ENST00000547954.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153553.hg.1 | NR_110048.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200153553.hg.1 | TC1200007588.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200153553.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153553.hg.1 | ENST00000546821.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153553.hg.1 | ENST00000547954.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200153553.hg.1 | NR_110048.1 | Refseq |
