Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008718.hg.1 |
ENST00000547465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008718.hg.1 |
ENST00000547465.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008718.hg.1 |
ENST00000548557.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008718.hg.1 |
ENST00000548557.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008718.hg.1 |
ENST00000548557.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008718.hg.1 |
ENST00000548557.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008718.hg.1 |
NR_110108.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008718.hg.1 |
NR_110108.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008718.hg.1 |
NR_110108.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008718.hg.1 |
NR_110108.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008718.hg.1 |
NR_110109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008718.hg.1 |
NR_110109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008718.hg.1 |
NR_110109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008718.hg.1 |
NR_110109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008718.hg.1 |
NR_110109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008718.hg.1 |
NR_110109.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008718.hg.1 |
NR_110110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008718.hg.1 |
NR_110110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008718.hg.1 |
NR_110110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008718.hg.1 |
NR_110110.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008718.hg.1 |
NR_110111.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008718.hg.1 |
NR_110111.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200008718.hg.1 |
NR_110111.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008718.hg.1 |
ENST00000547465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008718.hg.1 |
ENST00000547465.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008718.hg.1 |
ENST00000548557.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008718.hg.1 |
ENST00000548557.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008718.hg.1 |
ENST00000548557.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008718.hg.1 |
ENST00000548557.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200008718.hg.1 |
NR_110108.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008718.hg.1 |
NR_110108.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008718.hg.1 |
NR_110108.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008718.hg.1 |
NR_110108.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008718.hg.1 |
NR_110109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008718.hg.1 |
NR_110109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008718.hg.1 |
NR_110109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008718.hg.1 |
NR_110109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008718.hg.1 |
NR_110109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008718.hg.1 |
NR_110109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008718.hg.1 |
NR_110110.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008718.hg.1 |
NR_110110.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008718.hg.1 |
NR_110110.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008718.hg.1 |
NR_110110.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008718.hg.1 |
NR_110111.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008718.hg.1 |
NR_110111.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200008718.hg.1 |
NR_110111.2 |
Refseq |