Assay Details
Gene Symbol: CASC18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110109.1 | 3 - 4 | 170 | 139 | ||
GenBank mRNA | AB849960.1 | - | 3 - 4 | 170 | 139 | |
AB849961.1 | - | 3 - 4 | 170 | 139 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | chromosome 12 open reading frame 75 | C12orf75 |
AGD3, OCC-1, OCC1 |
-350.7kb | Sense | Chr. 12 - 105330636 - 105371518 on NCBI Build GRCh38 |
Downstream | NUAK family kinase 1 | NUAK1 | ARK5 | 326.0kb | Anti-sense | Chr. 12 - 106063347 - 106140033 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
