Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010566.hg.1 |
ENST00000606397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010566.hg.1 |
ENST00000606397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010566.hg.1 |
ENST00000606397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010566.hg.1 |
ENST00000606397.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010566.hg.1 |
ENST00000607680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010566.hg.1 |
ENST00000607680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010566.hg.1 |
ENST00000607680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010566.hg.1 |
ENST00000607680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010566.hg.1 |
ENST00000607680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010566.hg.1 |
ENST00000607680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010566.hg.1 |
NR_110383.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010566.hg.1 |
NR_110383.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010566.hg.1 |
NR_110383.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010566.hg.1 |
NR_110383.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010566.hg.1 |
NR_110383.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010566.hg.1 |
NR_110384.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010566.hg.1 |
NR_110384.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010566.hg.1 |
NR_110384.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171991.hg.1 |
ENST00000606397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171991.hg.1 |
ENST00000607680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171991.hg.1 |
ENST00000607680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171991.hg.1 |
NR_110383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171991.hg.1 |
NR_110384.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171991.hg.1 |
TC0X00010566.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00171991.hg.1 |
NR_110383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171991.hg.1 |
ENST00000606397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171991.hg.1 |
ENST00000607680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171991.hg.1 |
ENST00000607680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171991.hg.1 |
NR_110384.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171987.hg.1 |
ENST00000607680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171987.hg.1 |
ENST00000607680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171987.hg.1 |
NR_110383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171987.hg.1 |
NR_110384.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171987.hg.1 |
TC0X00010566.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00171987.hg.1 |
ENST00000607680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171987.hg.1 |
ENST00000607680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171987.hg.1 |
NR_110383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171987.hg.1 |
NR_110384.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171989.hg.1 |
ENST00000606397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171989.hg.1 |
NR_110383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171989.hg.1 |
TC0X00010566.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00171989.hg.1 |
ENST00000606397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171989.hg.1 |
NR_110383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010566.hg.1 |
ENST00000606397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010566.hg.1 |
ENST00000606397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010566.hg.1 |
ENST00000606397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010566.hg.1 |
ENST00000606397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010566.hg.1 |
ENST00000607680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010566.hg.1 |
ENST00000607680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010566.hg.1 |
ENST00000607680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010566.hg.1 |
ENST00000607680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010566.hg.1 |
ENST00000607680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010566.hg.1 |
ENST00000607680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010566.hg.1 |
NR_110383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010566.hg.1 |
NR_110383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010566.hg.1 |
NR_110383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010566.hg.1 |
NR_110383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010566.hg.1 |
NR_110383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010566.hg.1 |
NR_110384.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010566.hg.1 |
NR_110384.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010566.hg.1 |
NR_110384.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171990.hg.1 |
ENST00000606397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171990.hg.1 |
ENST00000607680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171990.hg.1 |
ENST00000607680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171990.hg.1 |
NR_110383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171990.hg.1 |
NR_110384.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171990.hg.1 |
TC0X00010566.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00171990.hg.1 |
ENST00000606397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171990.hg.1 |
ENST00000607680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171990.hg.1 |
ENST00000607680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171990.hg.1 |
NR_110383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171990.hg.1 |
NR_110384.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171988.hg.1 |
ENST00000606397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171988.hg.1 |
NR_110383.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171988.hg.1 |
TC0X00010566.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00171988.hg.1 |
ENST00000606397.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171988.hg.1 |
NR_110383.1 |
Refseq |