Assay Details
Gene Symbol: LOC388882 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110539.1 | 11 - 12 | 987 | 95 | ||
GenBank mRNA | BC036910.1 | - | 11 - 12 | 987 | 5164270 | 95 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101929374 | LOC101929374 | -27.0kb | Anti-sense | Chr. 22 - 23433564 - 23435071 on NCBI Build GRCh38 | |
Downstream | prostate cancer associated transcript 14 (non-protein coding) | PCAT14 | 70.8kb | Anti-sense | Chr. 22 - 23538102 - 23547306 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120569 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11443276 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11443300 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11443268 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11443307 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120680 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120581 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120694 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11443243 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120683 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120701 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120671 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120698 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120558 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17266660 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120640 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120585 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120675 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120706 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11443288 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17266580 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11443244 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120584 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120625 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11443245 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11443253 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120676 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120567 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11443296 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120707 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11443309 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120684 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11443293 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17266606 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120691 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11443256 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120619 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11443272 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11443308 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17266630 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11443279 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11443258 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120543 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17266566 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11443304 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120703 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120705 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17266649 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120556 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120644 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11443259 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11443292 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11443260 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11443297 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120690 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11443247 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120650 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17266686 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120607 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11443265 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120546 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11443261 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17266605 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120624 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11443262 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11443291 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17266685 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120565 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11443278 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P17266576 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120679 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P11443269 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P32120545 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 2327 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 48392 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P499005 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_32_P499005 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 1554940_PM_a_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200158423.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158423.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200158423.hg.1 | TC2200008169.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200158423.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200158423.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | ENST00000539460.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008169.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200158422.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158422.hg.1 | NR_110539.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200158422.hg.1 | TC2200008169.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200158422.hg.1 | ENST00000320372.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200158422.hg.1 | NR_110539.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 1554940_PM_a_at | - | - |
Human Genome U133 Plus 2.0 Array | 1554940_a_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554940_a_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554940_a_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554940_a_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554940_a_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554940_a_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554940_a_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554940_a_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554940_a_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554940_a_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554940_a_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1554940_a_at10 | - | - |
Human Genome U95 Set | 52299_PM_at | - | - |
Human Genome U95 Set | 52299_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1683149 | BC036910.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1792391 | - | - |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1683149 | BC036910.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1792391 | - | - |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1792391 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1683149 | BC036910.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1683149 | BC036910.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1683149 | BC036910.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1683149 | BC036910.1 | human_transcriptome |
