Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014725.hg.1 |
ENST00000418770.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014725.hg.1 |
ENST00000418770.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014725.hg.1 |
ENST00000418770.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014725.hg.1 |
ENST00000418770.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014725.hg.1 |
ENST00000418770.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014725.hg.1 |
ENST00000418770.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014725.hg.1 |
ENST00000418770.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014725.hg.1 |
ENST00000418770.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014725.hg.1 |
ENST00000453016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014725.hg.1 |
ENST00000453016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014725.hg.1 |
ENST00000453016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014725.hg.1 |
ENST00000453016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014725.hg.1 |
ENST00000453016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014725.hg.1 |
ENST00000453016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014725.hg.1 |
ENST00000453016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014725.hg.1 |
ENST00000453016.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014725.hg.1 |
ENST00000607836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014725.hg.1 |
ENST00000607836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014725.hg.1 |
ENST00000607836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014725.hg.1 |
ENST00000607836.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014725.hg.1 |
NR_110586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014725.hg.1 |
NR_110586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014725.hg.1 |
NR_110586.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014725.hg.1 |
NR_110586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200207105.hg.1 |
ENST00000418770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207105.hg.1 |
ENST00000418770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207105.hg.1 |
ENST00000453016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207105.hg.1 |
ENST00000453016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207105.hg.1 |
ENST00000607836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207105.hg.1 |
NR_110586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200207105.hg.1 |
TC0200014725.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200207105.hg.1 |
ENST00000418770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207105.hg.1 |
ENST00000418770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207105.hg.1 |
ENST00000453016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207105.hg.1 |
NR_110586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200207105.hg.1 |
ENST00000453016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207105.hg.1 |
ENST00000607836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014725.hg.1 |
ENST00000418770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014725.hg.1 |
ENST00000418770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014725.hg.1 |
ENST00000418770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014725.hg.1 |
ENST00000418770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014725.hg.1 |
ENST00000418770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014725.hg.1 |
ENST00000418770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014725.hg.1 |
ENST00000418770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014725.hg.1 |
ENST00000418770.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014725.hg.1 |
ENST00000453016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014725.hg.1 |
ENST00000453016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014725.hg.1 |
ENST00000453016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014725.hg.1 |
ENST00000453016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014725.hg.1 |
ENST00000453016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014725.hg.1 |
ENST00000453016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014725.hg.1 |
ENST00000453016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014725.hg.1 |
ENST00000453016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014725.hg.1 |
ENST00000607836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014725.hg.1 |
ENST00000607836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014725.hg.1 |
ENST00000607836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014725.hg.1 |
ENST00000607836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014725.hg.1 |
NR_110586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014725.hg.1 |
NR_110586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014725.hg.1 |
NR_110586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014725.hg.1 |
NR_110586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200207109.hg.1 |
ENST00000453016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207109.hg.1 |
ENST00000453016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207109.hg.1 |
ENST00000607836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207109.hg.1 |
NR_110586.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200207109.hg.1 |
TC0200014725.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200207109.hg.1 |
ENST00000453016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207109.hg.1 |
ENST00000453016.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207109.hg.1 |
ENST00000607836.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200207109.hg.1 |
NR_110586.1 |
Refseq |