Assay Details
Gene Symbol: LOC101929715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110597.1 | 4 - 5 | 756 | 77 | ||
GenBank mRNA | HY015289.1 | - | 3 - 4 | 425 | 77 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ornithine decarboxylase 1 | ODC1 | ODC | -5.3kb | Anti-sense | Chr. 2 - 10440371 - 10448554 on NCBI Build GRCh38 |
Downstream | nucleolar protein 10 | NOL10 | PQBP5 | 115.2kb | Anti-sense | Chr. 2 - 10570766 - 10689991 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
