Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010164.hg.1 |
ENST00000399581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010164.hg.1 |
ENST00000399581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010164.hg.1 |
ENST00000399581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010164.hg.1 |
ENST00000399581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010164.hg.1 |
ENST00000399581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010164.hg.1 |
ENST00000399581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010164.hg.1 |
ENST00000399581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010164.hg.1 |
ENST00000614414.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010164.hg.1 |
ENST00000614414.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010164.hg.1 |
ENST00000614414.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010164.hg.1 |
ENST00000614414.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010164.hg.1 |
ENST00000614414.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010164.hg.1 |
ENST00000614414.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010164.hg.1 |
ENST00000614414.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010164.hg.1 |
ENST00000614414.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010164.hg.1 |
NR_110646.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010164.hg.1 |
NR_110646.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010164.hg.1 |
NR_110646.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010164.hg.1 |
NR_110646.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010164.hg.1 |
NR_110646.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010164.hg.1 |
NR_110646.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010164.hg.1 |
NR_110646.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169337.hg.1 |
ENST00000399581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169337.hg.1 |
ENST00000614414.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169337.hg.1 |
NR_110646.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169337.hg.1 |
TC0X00010164.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00169337.hg.1 |
NR_110646.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169337.hg.1 |
ENST00000399581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169337.hg.1 |
ENST00000614414.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169343.hg.1 |
ENST00000614414.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169343.hg.1 |
NR_110646.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169343.hg.1 |
TC0X00010164.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00169343.hg.1 |
ENST00000614414.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169343.hg.1 |
NR_110646.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169350.hg.1 |
ENST00000614414.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169350.hg.1 |
NR_110646.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169350.hg.1 |
TC0X00010164.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00169350.hg.1 |
ENST00000614414.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169350.hg.1 |
NR_110646.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169341.hg.1 |
ENST00000399581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169341.hg.1 |
ENST00000614414.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169341.hg.1 |
NR_110646.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00169341.hg.1 |
TC0X00010164.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00169341.hg.1 |
ENST00000399581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169341.hg.1 |
ENST00000614414.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00169341.hg.1 |
NR_110646.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010164.hg.1 |
ENST00000399581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010164.hg.1 |
ENST00000399581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010164.hg.1 |
ENST00000399581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010164.hg.1 |
ENST00000399581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010164.hg.1 |
ENST00000399581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010164.hg.1 |
ENST00000399581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010164.hg.1 |
ENST00000399581.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010164.hg.1 |
ENST00000614414.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010164.hg.1 |
ENST00000614414.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010164.hg.1 |
ENST00000614414.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010164.hg.1 |
ENST00000614414.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010164.hg.1 |
ENST00000614414.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010164.hg.1 |
ENST00000614414.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010164.hg.1 |
ENST00000614414.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010164.hg.1 |
ENST00000614414.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010164.hg.1 |
NR_110646.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010164.hg.1 |
NR_110646.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010164.hg.1 |
NR_110646.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010164.hg.1 |
NR_110646.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010164.hg.1 |
NR_110646.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010164.hg.1 |
NR_110646.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010164.hg.1 |
NR_110646.1 |
Refseq |