Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009101.hg.1 |
ENST00000572353.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009101.hg.1 |
ENST00000572353.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009101.hg.1 |
ENST00000572353.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009101.hg.1 |
ENST00000574526.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009101.hg.1 |
ENST00000574526.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009101.hg.1 |
ENST00000574526.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009101.hg.1 |
NR_110851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009101.hg.1 |
NR_110851.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009101.hg.1 |
NR_110851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700168729.hg.1 |
ENST00000574526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168729.hg.1 |
NR_110851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700168729.hg.1 |
TC1700009101.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700168729.hg.1 |
NR_110851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700168729.hg.1 |
ENST00000574526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168727.hg.1 |
ENST00000572353.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168727.hg.1 |
ENST00000574526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168727.hg.1 |
NR_110851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700168727.hg.1 |
TC1700009101.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700168727.hg.1 |
ENST00000572353.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168727.hg.1 |
ENST00000574526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168727.hg.1 |
NR_110851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700168728.hg.1 |
ENST00000572353.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168728.hg.1 |
ENST00000574526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168728.hg.1 |
NR_110851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700168728.hg.1 |
TC1700009101.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700168728.hg.1 |
ENST00000572353.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168728.hg.1 |
ENST00000574526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700168728.hg.1 |
NR_110851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009101.hg.1 |
ENST00000572353.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009101.hg.1 |
ENST00000572353.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009101.hg.1 |
ENST00000572353.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009101.hg.1 |
ENST00000574526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009101.hg.1 |
ENST00000574526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009101.hg.1 |
ENST00000574526.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009101.hg.1 |
NR_110851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009101.hg.1 |
NR_110851.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009101.hg.1 |
NR_110851.1 |
Refseq |