Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008544.hg.1 |
ENST00000435946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008544.hg.1 |
ENST00000435946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008544.hg.1 |
ENST00000435946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008544.hg.1 |
ENST00000435946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008544.hg.1 |
ENST00000435946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008544.hg.1 |
NR_110856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008544.hg.1 |
NR_110856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008544.hg.1 |
NR_110856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008544.hg.1 |
NR_110856.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008544.hg.1 |
NR_110856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159438.hg.1 |
ENST00000435946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159438.hg.1 |
NR_110856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159438.hg.1 |
TC0600008544.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600159438.hg.1 |
ENST00000435946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159438.hg.1 |
NR_110856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159439.hg.1 |
ENST00000435946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159439.hg.1 |
NR_110856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159439.hg.1 |
TC0600008544.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600159439.hg.1 |
ENST00000435946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159439.hg.1 |
NR_110856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159434.hg.1 |
ENST00000435946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159434.hg.1 |
NR_110856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159434.hg.1 |
TC0600008544.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600159434.hg.1 |
ENST00000435946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159434.hg.1 |
NR_110856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159432.hg.1 |
ENST00000435946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159432.hg.1 |
NR_110856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159432.hg.1 |
TC0600008544.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600159432.hg.1 |
ENST00000435946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159432.hg.1 |
NR_110856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008544.hg.1 |
ENST00000435946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008544.hg.1 |
ENST00000435946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008544.hg.1 |
ENST00000435946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008544.hg.1 |
ENST00000435946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008544.hg.1 |
ENST00000435946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008544.hg.1 |
NR_110856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008544.hg.1 |
NR_110856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008544.hg.1 |
NR_110856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008544.hg.1 |
NR_110856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008544.hg.1 |
NR_110856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159440.hg.1 |
ENST00000435946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159440.hg.1 |
NR_110856.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159440.hg.1 |
TC0600008544.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600159440.hg.1 |
ENST00000435946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159440.hg.1 |
NR_110856.1 |
Refseq |