Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006914.hg.1 |
ENST00000572950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006914.hg.1 |
ENST00000572950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006914.hg.1 |
ENST00000572950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006914.hg.1 |
ENST00000572950.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006914.hg.1 |
NR_110907.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006914.hg.1 |
NR_110907.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006914.hg.1 |
NR_110907.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006914.hg.1 |
NR_110907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600149155.hg.1 |
ENST00000572950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600149155.hg.1 |
NR_110907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600149155.hg.1 |
TC1600006914.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600149155.hg.1 |
NR_110907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600149155.hg.1 |
ENST00000572950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006914.hg.1 |
ENST00000572950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006914.hg.1 |
ENST00000572950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006914.hg.1 |
ENST00000572950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006914.hg.1 |
ENST00000572950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006914.hg.1 |
NR_110907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006914.hg.1 |
NR_110907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006914.hg.1 |
NR_110907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006914.hg.1 |
NR_110907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600149154.hg.1 |
ENST00000572950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600149154.hg.1 |
NR_110907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600149154.hg.1 |
TC1600006914.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600149154.hg.1 |
ENST00000572950.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600149154.hg.1 |
NR_110907.1 |
Refseq |