Assay Details
Gene Symbol: LOC101927653 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120452.1 | 2 - 3 | 342 | 93 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | endogenous retrovirus group K member 8 Pol protein-like | LOC105369972 | -276.8kb | Sense | Chr. 12 - 58323535 - 58331743 on NCBI Build GRCh38 | |
Downstream | leucine rich repeats and immunoglobulin like domains 3 | LRIG3 | LIG3 | 255.0kb | Sense | Chr. 12 - 58869351 - 58920540 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
