Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000530896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000530896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000530896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000530896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000530896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000530896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000532217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000532217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000532217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
ENST00000532217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
NR_120571.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
NR_120571.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
NR_120571.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
NR_120571.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
NR_120571.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
NR_120571.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
NR_120571.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
NR_120571.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
NR_120571.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
NR_120571.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
NR_120571.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011782.hg.1 |
NR_120571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000530896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000530896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000530896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000530896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000530896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000530896.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000532217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000532217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000532217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
ENST00000532217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
NR_120571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
NR_120571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
NR_120571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
NR_120571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
NR_120571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
NR_120571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
NR_120571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
NR_120571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
NR_120571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
NR_120571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
NR_120571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011782.hg.1 |
NR_120571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100190988.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100190988.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100190988.hg.1 |
NR_120571.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100190988.hg.1 |
TC1100011782.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100190988.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100190988.hg.1 |
ENST00000500502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100190988.hg.1 |
NR_120571.1 |
Refseq |