Assay Details
Gene Symbol: MIR4300HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120571.1 | 6 - 7 | 947 | 84 | ||
GenBank mRNA | BC041900.1 | - | 6 - 7 | 947 | 84 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101928944 | LOC101928944 | -1.1mb | Sense | Chr. 11 - 80751200 - 80762803 on NCBI Build GRCh38 | |
Downstream | family with sequence similarity 181 member B | FAM181B | 716.3kb | Sense | Chr. 11 - 82732004 - 82733864 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000530896.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000530896.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000530896.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000530896.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000530896.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000530896.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000532217.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000532217.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000532217.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | ENST00000532217.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | NR_120571.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | NR_120571.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | NR_120571.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | NR_120571.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | NR_120571.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | NR_120571.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | NR_120571.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | NR_120571.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | NR_120571.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | NR_120571.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | NR_120571.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011782.hg.1 | NR_120571.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100190981.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100190981.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100190981.hg.1 | ENST00000530896.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100190981.hg.1 | ENST00000530896.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100190981.hg.1 | NR_120571.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100190981.hg.1 | TC1100011782.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100190981.hg.1 | ENST00000530896.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100190981.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100190981.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100190981.hg.1 | ENST00000530896.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100190981.hg.1 | NR_120571.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100190982.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100190982.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100190982.hg.1 | NR_120571.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100190982.hg.1 | TC1100011782.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100190982.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100190982.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100190982.hg.1 | NR_120571.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000500502.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000530896.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000530896.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000530896.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000530896.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000530896.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000530896.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000532217.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000532217.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000532217.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | ENST00000532217.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | NR_120571.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | NR_120571.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | NR_120571.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | NR_120571.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | NR_120571.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | NR_120571.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | NR_120571.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | NR_120571.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | NR_120571.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | NR_120571.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | NR_120571.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011782.hg.1 | NR_120571.1 | Refseq |
