Assay Details
Gene Symbol: LINC01495 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120583.1 | 2 - 3 | 247 | 113 | ||
GenBank mRNA | BX092575.1 | - | 2 - 3 | 246 | 113 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | solute carrier family 17 member 6 | SLC17A6 |
DNPI, VGLUT2 |
-88.3kb | Anti-sense | Chr. 11 - 22338121 - 22379503 on NCBI Build GRCh38 |
Downstream | Fanconi anemia complementation group F | FANCF | FAF | 142.3kb | Sense | Chr. 11 - 22622533 - 22625841 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
