Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011116.hg.1 |
ENST00000415302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011116.hg.1 |
ENST00000415302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011116.hg.1 |
ENST00000415302.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011116.hg.1 |
ENST00000423523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011116.hg.1 |
ENST00000423523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011116.hg.1 |
ENST00000423523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011116.hg.1 |
ENST00000423523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011116.hg.1 |
NR_121581.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011116.hg.1 |
NR_121581.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011116.hg.1 |
NR_121581.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011116.hg.1 |
NR_121581.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011116.hg.1 |
NR_121581.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011116.hg.1 |
NR_121581.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011116.hg.1 |
NR_121581.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011116.hg.1 |
NR_121581.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011116.hg.1 |
NR_121581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011116.hg.1 |
ENST00000415302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011116.hg.1 |
ENST00000415302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011116.hg.1 |
ENST00000415302.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011116.hg.1 |
ENST00000423523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011116.hg.1 |
ENST00000423523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011116.hg.1 |
ENST00000423523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011116.hg.1 |
ENST00000423523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011116.hg.1 |
NR_121581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011116.hg.1 |
NR_121581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011116.hg.1 |
NR_121581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011116.hg.1 |
NR_121581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011116.hg.1 |
NR_121581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011116.hg.1 |
NR_121581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011116.hg.1 |
NR_121581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011116.hg.1 |
NR_121581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011116.hg.1 |
NR_121581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900175637.hg.1 |
ENST00000423523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900175637.hg.1 |
NR_121581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900175637.hg.1 |
TC0900011116.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900175637.hg.1 |
NR_121581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900175637.hg.1 |
ENST00000423523.1 |
Ensembl |