Assay Details
Gene Symbol: FOXD3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_121635.1 | 1 - 2 | 147 | 141 | ||
GenBank mRNA | KF925351.1 | - | 1 - 2 | 147 | 141 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 466 | LINC00466 | -4.1kb | Sense | Chr. 1 - 63159083 - 63317257 on NCBI Build GRCh38 | |
Downstream | forkhead box D3 | FOXD3 |
AIS1, Genesis, HFH2, VAMAS2 |
601.0 | Anti-sense | Chr. 1 - 63323059 - 63325126 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
