Assay Details
Gene Symbol: FOXD3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_121637.1 | 1 - 2 | 87 | 90 | ||
GenBank mRNA | KF925349.1 | - | 1 - 2 | 87 | 90 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 466 | LINC00466 | -4.1kb | Sense | Chr. 1 - 63159083 - 63317257 on NCBI Build GRCh38 | |
Downstream | microRNA 6068 | MIR6068 | hsa-mir-6068 | 2.5kb | Sense | Chr. 1 - 63326925 - 63326984 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121634.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121634.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121634.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121634.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121634.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121634.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121635.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121635.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121635.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121635.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121635.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121636.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121636.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121636.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121636.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121636.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121637.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121637.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121637.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | NR_121637.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000427268.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000427268.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000427268.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000427268.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000449386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121634.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121634.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121634.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121634.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121634.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121634.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121635.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121635.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121635.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121635.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121635.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121636.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121636.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121636.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121636.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121636.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121637.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121637.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121637.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | NR_121637.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000418244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000426393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000427268.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000427268.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000427268.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000427268.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100014420.hg.1 | ENST00000431294.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207994.hg.1 | ENST00000427268.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207994.hg.1 | NR_121637.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100207994.hg.1 | TC0100014420.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100207994.hg.1 | ENST00000427268.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100207994.hg.1 | NR_121637.1 | Refseq |
