Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
ENST00000504332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
ENST00000504332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
ENST00000504332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
ENST00000504332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
ENST00000504332.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
NR_125384.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
NR_125384.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
NR_125384.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
NR_125384.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
NR_125384.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
NR_125384.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
NR_125385.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
NR_125385.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
NR_125385.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
NR_125385.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010419.hg.1 |
NR_125385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
ENST00000504332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
ENST00000504332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
ENST00000504332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
ENST00000504332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
ENST00000504332.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
NR_125384.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
NR_125384.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
NR_125384.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
NR_125384.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
NR_125384.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
NR_125384.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
NR_125385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
NR_125385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
NR_125385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
NR_125385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010419.hg.1 |
NR_125385.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400177036.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400177036.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400177036.hg.1 |
NR_125384.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400177036.hg.1 |
TC1400010419.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1400177036.hg.1 |
NR_125384.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400177036.hg.1 |
ENST00000551271.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400177036.hg.1 |
ENST00000551271.1 |
Ensembl |