Assay Details
Gene Symbol: LOC100507437 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125385.1 | 1 - 2 | 722 | 95 | ||
GenBank mRNA | AK123017.1 | - | 1 - 2 | 95 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | testis expressed 22 | TEX22 | -3.7kb | Anti-sense | Chr. 14 - 105398499 - 105413859 on NCBI Build GRCh38 | |
Downstream | metastasis associated 1 | MTA1 | 110.0 | Anti-sense | Chr. 14 - 105419849 - 105470729 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | ENST00000504332.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | ENST00000504332.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | ENST00000504332.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | ENST00000504332.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | ENST00000504332.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | NR_125384.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | NR_125384.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | NR_125384.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | NR_125384.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | NR_125384.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | NR_125384.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | NR_125385.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | NR_125385.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | NR_125385.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | NR_125385.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010419.hg.1 | NR_125385.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400177040.hg.1 | ENST00000504332.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177040.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177040.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177040.hg.1 | NR_125384.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400177040.hg.1 | NR_125385.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400177040.hg.1 | TC1400010419.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400177040.hg.1 | NR_125384.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400177040.hg.1 | ENST00000504332.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177040.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177040.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177040.hg.1 | NR_125385.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400177041.hg.1 | ENST00000504332.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177041.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177041.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177041.hg.1 | NR_125384.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400177041.hg.1 | NR_125385.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400177041.hg.1 | TC1400010419.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400177041.hg.1 | ENST00000504332.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177041.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177041.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177041.hg.1 | NR_125384.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400177041.hg.1 | NR_125385.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400177033.hg.1 | ENST00000504332.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177033.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177033.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177033.hg.1 | NR_125384.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400177033.hg.1 | NR_125385.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400177033.hg.1 | TC1400010419.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400177033.hg.1 | ENST00000504332.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177033.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177033.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177033.hg.1 | NR_125384.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400177033.hg.1 | NR_125385.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400177039.hg.1 | ENST00000504332.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177039.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177039.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177039.hg.1 | NR_125384.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400177039.hg.1 | NR_125385.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400177039.hg.1 | TC1400010419.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400177039.hg.1 | ENST00000504332.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177039.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177039.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177039.hg.1 | NR_125384.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400177039.hg.1 | NR_125385.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400177034.hg.1 | ENST00000504332.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177034.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177034.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177034.hg.1 | NR_125384.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400177034.hg.1 | NR_125385.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400177034.hg.1 | TC1400010419.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400177034.hg.1 | ENST00000504332.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177034.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177034.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400177034.hg.1 | NR_125384.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400177034.hg.1 | NR_125385.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | ENST00000504332.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | ENST00000504332.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | ENST00000504332.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | ENST00000504332.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | ENST00000504332.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | ENST00000551271.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | NR_125384.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | NR_125384.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | NR_125384.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | NR_125384.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | NR_125384.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | NR_125384.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | NR_125385.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | NR_125385.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | NR_125385.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | NR_125385.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010419.hg.1 | NR_125385.1 | Refseq |
