Assay Details
Gene Symbol: LOC101928540 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125859.1 | 1 - 2 | 38 | 75 | ||
GenBank mRNA | HY214668.1 | - | 1 - 2 | 76 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC100506804 | LOC100506804 | -91.3kb | Sense | Chr. 6 - 75285014 - 75291864 on NCBI Build GRCh38 | |
Downstream | microRNA 4463 | MIR4463 | 42.7kb | Sense | Chr. 6 - 75428407 - 75428473 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
