Assay Details
Gene Symbol: LOC101928540 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125859.1 | 5 - 6 | 303 | 101 | ||
GenBank mRNA | DB456186.1 | - | 4 - 5 | 299 | 101 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 4463 | MIR4463 | -24.5kb | Sense | Chr. 6 - 75428407 - 75428473 on NCBI Build GRCh38 | |
Downstream | SUMO1/sentrin specific peptidase 6 | SENP6 |
SSP1, SUSP1 |
146.5kb | Sense | Chr. 6 - 75601509 - 75718285 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
