Assay Details
Gene Symbol: LOC102723883 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125869.1 | 1 - 2 | 85 | 97 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | solute carrier family 25 member 51 pseudogene 1 | SLC25A51P1 |
MCART3P, bA707M13.1 |
-2.3mb | Sense | Chr. 6 - 65787879 - 65789483 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC101928280 | LOC101928280 | 166.5kb | Anti-sense | Chr. 6 - 68226970 - 68329899 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
