Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007963.hg.1 |
ENST00000505175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007963.hg.1 |
ENST00000505175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007963.hg.1 |
ENST00000505175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007963.hg.1 |
ENST00000505175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007963.hg.1 |
ENST00000505175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007963.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007963.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007963.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007963.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007963.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007963.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007963.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007963.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007963.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007963.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007963.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007963.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007963.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007963.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007963.hg.1 |
NR_125908.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007963.hg.1 |
NR_125908.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007963.hg.1 |
NR_125908.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007963.hg.1 |
NR_125908.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007963.hg.1 |
NR_125908.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007963.hg.1 |
NR_125908.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007963.hg.1 |
NR_125908.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154850.hg.1 |
ENST00000505175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154850.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154850.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154850.hg.1 |
NR_125908.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154850.hg.1 |
TC0400007963.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400154850.hg.1 |
NR_125908.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154850.hg.1 |
ENST00000505175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154850.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154850.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154849.hg.1 |
ENST00000505175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154849.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154849.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154849.hg.1 |
NR_125908.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400154849.hg.1 |
TC0400007963.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400154849.hg.1 |
ENST00000505175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154849.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154849.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400154849.hg.1 |
NR_125908.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007963.hg.1 |
ENST00000505175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007963.hg.1 |
ENST00000505175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007963.hg.1 |
ENST00000505175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007963.hg.1 |
ENST00000505175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007963.hg.1 |
ENST00000505175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007963.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007963.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007963.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007963.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007963.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007963.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007963.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007963.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007963.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007963.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007963.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007963.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007963.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007963.hg.1 |
ENST00000512502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007963.hg.1 |
NR_125908.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007963.hg.1 |
NR_125908.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007963.hg.1 |
NR_125908.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007963.hg.1 |
NR_125908.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007963.hg.1 |
NR_125908.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007963.hg.1 |
NR_125908.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007963.hg.1 |
NR_125908.1 |
Refseq |